Pyranine ≥98% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-H0034-G001 CDX-H0034-G005 CDX-H0034-G010
102990-224EA 157.85 USD
102990-224 102990-226 102990-228
Pyranine ≥98% (by HPCE)
Pyranine

Useful for optical sensor applications. Fluorescent pH indicator for physiological range.

Formula: C₁₆H₇Na₃O₁₀S₃
MW: 524.371 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037575
CAS Number: 6358-69-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPCE)
Appearance/Color Yellow powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR