FLUO 3/AM ≥90% (by TLC)

Supplier: ADIPOGEN CORP MS

CDX-F0033-M001 CDX-F0033-M010
102989-956EA 342 USD
102989-956 102989-958
FLUO 3/AM ≥90% (by TLC)
FLUO 3/AM

Ca2+-indicator fluo-3 for use with visible-light excitation sources in flow cytometry and confocal laser-scanning microscopy. Since about 1996, fluo-3 has also been extensively used in cell-based high-throughput screening assays for drug discovery. Fluo-3 AM is essentially nonfluorescent until it is hydrolyzed intracellularly and/or in the presence of Ca2+. The green-fluorescent emission (~525 nm) of Ca2+-bound fluo-3 is conventionally detected using optical filter sets designed for fluorescein (FITC). As prepared by Minta, Kao and Tsien, fluo-3 was originally reported to undergo an ~40-fold increase in fluorescence upon binding Ca2+.

Formula: C₅₁H₅₀Cl₂N₂O₂₃
Storage Temperature: Freezer
CAS Number: 121714-22-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (TLC)
Appearance/Color Dark red powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR