Enalaprilat ≥99% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
(S)-1-((S)-2-(((S)-1-Carboxy-3-phenylpropyl)amino)propanoyl)pyrrolidine-2-carboxylic acid
CDX-E0090-M050
CDX-E0090-M250
102989-928EA
210.46
USD
102989-928
102989-930
Enalaprilat ≥99% (by HPLC)
Enalaprilat
Active metabolite enalapril. Nonsulfhydryl angiotensin-converting enzyme (ACE) inhibitor. Shown to improve cardiac preservation via bradykinin receptor and PKC without affecting ATP metabolism. Has some antioxidant activity, which does not include the PKC-NADPH oxidase pathway. Increases PPARbeta/delta expression, without influence on PPARalpha and PPARgamma.
Formula:
C₁₈H₂₈N₂O₇ MW: 384.42 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00865786 CAS Number: 76420-72-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Slightly soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...