Epoxy soybean oil ≥98%
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-E0037-G010
CDX-E0037-G050
102989-900EA
157.85
USD
102989-900
102989-902
Epoxy soybean oil ≥98%
Soybean oil, epoxidised
Useful for the determination of ESBO in foods with HPLC-MS or GC-MS. ESBO is currently employed as plasticizer and HCl scavenger in PVC products. Vegetable oils are widely used for chemical manipulation because of the high numbers of carbon-carbon double bonds. Through the epoxidation, an epoxide group (which is a more reactive group than a double bond) is added to the soybean oil, transforming it in a good hydrochloric acid scavenger and good plasticizer.
Formula:
C₅₇H₉₈O₁₂ MW: 975.399 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00163560 CAS Number: 8013-07-8 UN: 3082 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Yellow to brown oil. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...