DMNQ ≥99% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
2,3-Dimethoxy-1,4-naphthoquinone, 2,3-Dimethoxynaphthalene-1,4-dione
CDX-D0205-M010
CDX-D0205-M050
102989-664EA
210.46
USD
102989-664
102989-666
DMNQ ≥99% (by HPLC)
DMNQ
A cell-permeable, non-alkylating, non-thiol, adduct-forming, redox cycling quinone. Generates superoxide production and induces cell proliferation, apoptosis and necrosis in vitro. Redox cycling agent.
Formula:
C₁₂H₁₀O₄ MW: 218.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00184331 CAS Number: 6956-96-3 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Yellow powder. |
Solubility | Soluble in DMSO or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...