DAF-2 diacetate ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-D0085-M001 CDX-D0085-M005 CDX-D0085-M010
102987-770EA 368.31 USD
102987-770 102987-772 102987-774
DAF-2 diacetate ≥95% (by HPLC)
DAF-2 diacetate

DAF-2 DA is probably the most successful indicator for nitric oxide. As with other fluorescein diacetates, DAF-2 diacetate is membrane permeant and is deacetylated by intracellular esterases to 4,5-diaminofluorescein (DAF-2).


Can be used for the fluorimetric detection of nitric oxide and in fluorescence microscopy to measure real-time changes in nitric oxide levels in vivo. Spectral Data: lambdaex 491nm; lambdaem 513nm.

Formula: C₂₄H₁₈N₂O₇
MW: 446.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05865101
CAS Number: 205391-02-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Off-white powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR