DAF-2 ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0084-M001 CDX-D0084-M005 CDX-D0084-M010
102987-764EA 368.31 USD
102987-764 102987-766 102987-768
DAF-2 ≥95% (by HPLC)
DAF-2

DAF-2 is highly sensitive reagent for NO detection and determination of nitric oxide synthase activity. DAF-2, however, remains essentially nonfluorescent until it reacts with the nitrosonium cation (produced by spontaneous oxidation of nitric oxide) to form a fluorescent heterocycle, which becomes trapped in the cell's cytoplasm. This sensitive fluorescent probe has been used to identify individual nitric oxide-producing neurons in brain slices, in mitochondria and in living plant cells. Spectral data: lambdaex 505nm; lambdaem 518nm in 0.1M Tris pH8.0.

Formula: C₂₀H₁₆N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD01866182
CAS Number: 205391-01-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR