(+)-1-Deoxynojirimycin ≥98%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
1-Deoxynojirimycin, (2R,3R,4R,5S)-2-(Hydroxymethyl)piperidine-3,4,5-triol
BVT-0112-M001
BVT-0112-M005
102988-892EA
27.69
USD
102988-892
102988-894
(+)-1-Deoxynojirimycin ≥98%
(+)-1-Deoxynojirimycin
Polyhydroxylated piperidine alkaloid. alpha-Glucosidase I and II inhibitor. Antidiabetic. Antihyperglycaemic activity. Animal feedstuff additive. Antiviral. Shows anti-HIV activity.
Formula:
C₆H₁₃NO₄ MW: 163.17 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00063474 CAS Number: 19130-96-2 |
Specification Test Results
Source/Host | Isolated from Streptomyces sp. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO (10mg/ml) or water (10mg/ml). |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...