(S)-CR8 99
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-MR-C0004-M010
AG-MR-C0004-M050
102987-166EA
360
USD
102987-166
102987-168
(S)-CR8 99
(S)-CR8
Potent cyclin-dependent kinase (CDK) 1, 2, 5, and 9 inhibitor. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Inhibits cysts formation in culture. Casein kinase 1 (CK1delta/epsilon) and Dyrk1A inhibitor. Down regulates expression of Mcl-1 and MYCN. Neuroprotective in experimental traumatic brain injury. Potential anti-inflammatory compound. Potential antidiabetic compound.
Formula:
C₂₄H₂₉N₇O MW: 431.5 g/mol Storage Temperature: Freezer |
CAS Number:
1084893-56-0 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...