(R)-CR8 98

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (R)-2-((9-Isopropyl-6-((4-(pyridin-2-yl)benzyl)amino)-9H-purin-2-yl)amino)butan-1-ol

AG-MR-C0003-M010 AG-MR-C0003-M050
102987-466EA 360 USD
102987-466 102987-468
(R)-CR8 98
(R)-CR8

Potent and selective inhibitor of cyclin dependent kinases CDK1, CDK2, CDK5, CDK7 and CDK9. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Casein kinase 1 (CK1delta/epsilon) and glycogen synthase kinase (GSK-3alpha/beta) inhibitor. Potential anti-inflammatory compound. Potential antidiabetic compound.

Formula: C₂₄H₂₉N₇O
MW: 431.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18785641
CAS Number: 294646-77-8

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR