Monosodium urate

Supplier: ADIPOGEN CORP MS

Synonyms: Sodium 2,6,8-trioxo-1,2,6,7,8,9-hexahydropurin-3-ide, Uric acid sodium, MSU, Uric acid

AG-CR1-3950-M002 AG-CR1-3950-2002
102516-336EA 116.31 USD
102516-336 102987-444
Monosodium urate
Monosodium urate

NLRP3/NALP3 inflammasome activator. Specially crystallized and tested for biological activity.
Stimulates the caspase-1 activating NLRP3/NALP3 inflammasome, resulting in the production of active IL-1beta and IL-18.


Appearance: Solid crystals. Solution may appear as a slightly hazy suspension.
Solubility: Not soluble in PBS. Reconstitute 2mg of MSU in 100μl PBS to obtain a 20mg/ml suspension.
Endotoxin: Content <0.001EU/μg purified protein (LAL test; Lonza)

Short Term Storage: +4°C
Long Term Storage: -20°C
Handling Advice: Vortex before use.

Formula: C₅H₃N₄O₃·Na
MW: 190.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067310
CAS Number: 1198-77-2

Order Now

Specification Test Results

Appearance/Color Solid crystals. Solution may appear as a slightly hazy suspension.
Solubility Not soluble in PBS. Reconstitute 2mg of MSU in 100µl PBS to obtain a 20mg/ml suspension.
0.001EU/µg purified protein (LAL test; Lonza).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR