Bortezomib ≥97% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
((R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butyl)boronic acid
AG-CR1-3602-M005
AG-CR1-3602-M025
102988-732EA
76.15
USD
102988-732
102988-734
Bortezomib ≥97% (by NMR)
Bortezomib
Highly potent, selective and reversible cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Anticancer compound. Inhibits proliferation and migration of several tumor cell lines with nanomolar potency. Apoptosis inducer.
Formula:
C₁₉H₂₅BN₄O₄ MW: 384.24 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09056737 CAS Number: 179324-69-7 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO, ethanol or methanol. Slightly soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...