DMNQ ≥99% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2,3-Dimethoxy-1,4-naphthoquinone, 2,3-Dimethoxynaphthalene-1,4-dione

AG-CR1-3598-M005 AG-CR1-3598-M010 AG-CR1-3598-M025
102988-720EA 96.92 USD
102988-720 102988-722 102988-724
DMNQ ≥99% (by NMR)
DMNQ

Cell permeable, non-alkylating, non-thiol, adduct-forming, redox cycling quinone. Intracellular superoxide anion formation/ROS generation inducer. Anticancer agent. Shown to induce cell proliferation, apoptosis, necrosis and necroptosis in vitro, dependent on concentration, time, temperature and cell type. Valuable tool for the generation of reactive oxygen species (ROS) in order to study the role of ROS in cell toxicity, apoptosis and necrosis. Useful as reference compound in characterizing the effects of oxidative stress. Can be used to eliminate any mechanistic ambiguity involving redox cycling quinoids as the source of reactive oxidant species/oxidative stress in biological studies.

Formula: C₁₂H₁₀O₄
MW: 218.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00184331
CAS Number: 6956-96-3

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color Yellow crystalline solid.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR