4-Aminophenylphosphate sodium salt ≥97%
Supplier: ADIPOGEN CORP MS
Synonyms:
4-APP
AG-CR1-3586-M010
AG-CR1-3586-M050
AG-CR1-3586-M100
102988-658EA
34.62
USD
102988-658
102988-660
102988-662
4-Aminophenylphosphate sodium salt ≥97%
4-Aminophenylphosphate sodium salt
Substrate for the electrochemical measurement of alkaline phosphatase.
Formula:
C₆H₇NO₄P·Na·4.0H₂O Storage Temperature: Freezer |
CAS Number:
108084-47-5 |
Specification Test Results
Purity |
|
Appearance/Color | Off-white to light gray crystalline solid. |
Solubility | Soluble in methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...