MTSSL ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: (1-Oxyl-2,2,5,5-tetramethylpyrroline-3-methyl)methanethiosulphonate

AG-CR1-3573-M010 AG-CR1-3573-M050
102988-600EA 110.77 USD
102988-600 102988-602
MTSSL ≥98%
MTSSL

Highly reactive, thiol-specific spin-label. Specific conformational probe of thiol site structure by its minimal rotational freedom and distance from the covalent disulfide linkage to the macromolecule under study. Used to label cysteine residues in proteins (site-directed labeling, SDS-labeling). Allows protein structure and protein dynamics determination as well as the study of protein-protein and protein-oligonucleotide interactions.

Formula: C₁₀H₁₈NO₃S₂
MW: 264.3 g/mol
Storage Temperature: Freezer
CAS Number: 81213-52-7

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color Yellow crystalline solid.
Solubility Soluble in ethanol, methanol, DMSO, acetonitrile or acetone. *See below for solubility in aqueous solutions.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR