Synthetic Leupeptin Hemisulfate (from Synthetic)
Supplier: ADIPOGEN CORP MS
AG-CP3-7000-M005 AG-CP3-7000-M025 AG-CP3-7000-M100
102988-512EA 48.46 USD
102988-512 102988-514 102988-516
Synthetic Leupeptin Hemisulfate (from Synthetic)
Proteins and Peptides
Potent, competitive and reversible cysteine, serine and threonine protease inhibitor. Inhibits calpain, kallikrein, trypsin, plasmin, papain and cathepsin B. Does not inhibit pepsin, elastase, renin, cathepsins A and D, thrombin or alpha-chymotrypsin. Used to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. Shown to inhibit activation-induced programmed cell death. Antioxidant and anti-inflammatory agent. Widely used as a protein purification tool to prevent proteases present in tissue samples from degrading the protein of interest.
  • White to off-white powder
  • Soluble in ethanol, methanol or water