Asiaticoside ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: (1S,2R,4aS,6aS,6bR,8aR,9R,10R,11R,12aR,12bR,14bS)-(2S,3R,4S,5S,6R)-6-((((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl 10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate

AG-CN2-0080-M010 AG-CN2-0080-M050
102987-276EA 83.08 USD
102987-276 102987-278
Asiaticoside ≥95% (by NMR)
Asiaticoside

Antibacterial. Human collagen I synthesis inducer. Anti-wrinkle activity. Shows wound healing activity. Enhances normal human skin cell migration, attachment and growth. Suppresses collagen expression and TGF-beta/Smad signaling by inducing Smad7 and inhibiting TGF-betaRI and TGF-betaRII. Anti-inflammatory. Anti-gastric ulcer. Mitogen-activated protein kinases (MAPKs), nuclear factor-kappaB (NF-kappaB), cyclooxygenase-2 (COX-2) and inducible nitric oxide synthase (iNOS; NOSII) expression, production of serum tumor necrosis factor (TNF-alpha) and interleukin-6 (IL-6) inhibitor. Apoptosis inducer in cancer cells. Enhances antitumor activity of vincristine in cancer cells. Might be useful in cancer chemotherapy. Anxiolytic. Antidepressant-like activity. Hepatoprotective effects. CYP2C19 and CYP3A4 inhibitor. Neuroprotective. Antileishmanial activity. cPLA2 and sPLA2 inhibitor.

Formula: C₄₈H₇₈O₁₉
MW: 959.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06642601
CAS Number: 16830-15-2

Order Now

Specification Test Results

Source/Host Isolated from Centella asiatica.
Purity 95% (NMR)
Appearance/Color White solid.
Solubility Soluble in ethanol, methanol or pyridine. Insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR