Wogonin ≥98%
Supplier: ADIPOGEN CORP MS
Synonyms:
5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone
Anti-inflammatory. Increases nitric oxide (NO) production. Inhibits PGE2, cyclooxygenase-2 (COX-2) and inducible nitric oxide synthase (iNOS; NOS II). Antioxidant. Anti-viral. Shows anti-hepatitis B virus activity. MCP-1 inhibitor. Shows anxiolytic effect through modulation of the GABA(A) receptor complex. Antifungal. Neuroprotective. Anticancer compound. Apoptosis and cell cycle arrest inducer. Sensitizes TNFalpha- and TRAIL-induced apoptosis. Antiangiogenic. NF-kappaB inhibitor. Telomerase activity inhibitor. CDK9 inhibitor. PI3K-AKT pathway modulator. Autophagy inducer.
Formula:
C₁₆H₁₂O₅ MW: 284.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00017736 CAS Number: 632-85-9 |
Specification Test Results
Source/Host | Isolated from Scutellaria baicalensis Georgi. |
Purity |
|
Appearance/Color | Yellow solid. |
Solubility | Soluble in DMSO and methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...