ER 172594-06 hydrobromide ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
SYN-3002-M001
SYN-3002-M005
SYN-3002-M010
102992-840EA
292.15
USD
102992-840
102992-842
102992-844
ER 172594-06 hydrobromide ≥95%
ER 172594-06 hydrobromide
E5555 shows potent inhibitory effects on human platelet aggregation induced by thrombin and TRAP with IC values of 0.064 and 0.031µM, respectively.
Formula:
C₂₉H₃₉BrFN₃O₅·HBr Storage Temperature: Freezer |
CAS Number:
474550-69-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...