AST-487 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
1-(4-((4-Ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-3-(4-((6-(methylamino)pyrimidin-4-yl)oxy)phenyl)urea
SYN-1210-M001
SYN-1210-M005
SYN-1210-M010
SYN-1210-M050
102992-804EA
134.31
USD
102992-804
102992-806
102992-808
102992-810
AST-487 ≥95%
AST-487
AST 487 displays high selectivity and potency toward FLT3 as a molecular target, and which could potentially be used to override drug resistance in AML.
Formula:
C₂₆H₃₀F₃N₇O₂ MW: 529.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11983171 CAS Number: 630124-46-8 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...