PRT-060318 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(((1R,2S)-2-Aminocyclohexyl)amino)-4-(m-tolylamino)pyrimidine-5-carboxamide

SYN-1204-M001 SYN-1204-M005 SYN-1204-M010
102992-778EA 224.31 USD
102992-778 102992-780 102992-782
PRT-060318 ≥95%
PRT-060318

PRT-060318, a novel Syk inhibitor, prevents heparin-induced thrombocytopenia and thrombosis in a transgenic mouse model.

Formula: C₁₈H₂₄N₆O
MW: 340.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28155088
CAS Number: 1194961-19-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR