GDC-0032 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
Taselisib, 2-(4-(2-(1-Isopropyl-3-methyl-1H-1,2,4-triazol-5-yl)-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepin-9-yl)-1H-pyrazol-1-yl)-2-methylpropanamide
SYN-1202-M001
SYN-1202-M005
SYN-1202-M010
SYN-1202-M050
102992-762EA
119.08
USD
102992-762
102992-764
102992-766
102992-768
GDC-0032 ≥95%
GDC-0032
GDC-0032 is a potent, next-generation beta isoform-sparing PI3K inhibitor targeting PI3Kalpha/delta/gamma with Ki of 0.29nM/0.12nM/0.97nM,
Formula:
C₂₄H₂₈N₈O₂ MW: 460.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD28167893 CAS Number: 1282512-48-4 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...