BKM120 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: (5-[2,6-Di(4-morpholinyl)-4-pyrimidinyl]-4-(trifluoromethyl)-2-pyridinamine, 5-(2,6-Dimorpholinopyrimidin-4-yl)-4-(trifluoromethyl)pyridin-2-amine, Buparlisib, NVP-BKM120

SYN-1200-M001 SYN-1200-M005 SYN-1200-M010 SYN-1200-M050
102992-746EA 90 USD
102992-746 102992-748 102992-750 102992-752
BKM120 ≥95%
BKM120

NVP-BKM120 is a potent, selective, orally bioavailable Class I PI3Kalpha Inhibitor with IC(50) of 20nM, currently in Phase II clinical trials for the treatment of cancer. It exhibits lower potency against class III and class IV PI3Ks.

Formula: C₁₈H₂₁F₃N₆O₂
MW: 410.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18251596
CAS Number: 944396-07-0

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR