LDK378 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
LDK-378
, 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamine
, Ceritinib (free base)
, 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulphonyl)phenyl)pyrimidine-2,4-diamine
SYN-1199-M010
SYN-1199-M050
SYN-1199-M001
SYN-1199-M005
102992-744EA
582.92
USD
102992-744
102992-738
102992-742
102992-740
LDK378 ≥95%
LDK378
LDK378 is potent inhibitor against ALK with IC(50) of 0.2nM, shows 40- and 35-fold selectivity against IGF-1R and InsR, respectively.
Formula:
C₂₈H₃₆ClN₅O₃S MW: 558.1 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD26142648 CAS Number: 1032900-25-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO (heat) or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...