GDC-046 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide
SYN-1198-M001
SYN-1198-M005
SYN-1198-M010
SYN-1198-M050
102992-730EA
134.31
USD
102992-730
102992-732
102992-734
102992-736
GDC-046 ≥95%
GDC-046
GDC046 is a potent selective TYK2 inhibitor (Ki=4.8nm) versus JAK1 (Ki=84nm) and JAK2 (Ki=28nm). It is a lead analog with good kinase selectivity, physicochemical properties, and pharmacokinetic profile.
Formula:
C₁₆H₁₃Cl₂N₃O₂ MW: 350.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD28155087 CAS Number: 1258292-64-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...