BLZ-945 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: BLZ945, Sotuletinib, 4-((2-(((1R,2R)-2-Hydroxycyclohexyl)amino)benzo[d]thiazol-6-yl)oxy)-N-methylpicolinamide

SYN-1197-M001 SYN-1197-M005 SYN-1197-M010
102992-724EA 239.54 USD
102992-724 102992-726 102992-728
BLZ-945 ≥95%
BLZ-945

BLZ945 is an orally active, potent and selective colony stimulating factor 1 receptor (CSF-1R) inhibitor, which inhibits CSF-1R activity with an IC(50) of 1nM and is more than 1000-fold selective against its closest receptor tyrosine kinase homologs c-KIT and Platelet-derived Growth Factor Receptor beta (PDGFRbeta ) as well as more than 200 additional kinases, confirming the selectivity of the compound.

Formula: C₂₀H₂₂N₄O₃S
MW: 398.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28142668
CAS Number: 953769-46-5

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR