LY2784544 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
3-(4-Chloro-2-fluorobenzyl)-2-methyl-N-(5-methyl-1H-pyrazol-3-yl)-8-(morpholinomethyl)imidazo[1,2-b]pyridazin-6-amine, Gandotinib
SYN-1196-M001
SYN-1196-M005
SYN-1196-M010
SYN-1196-M050
102992-716EA
134.31
USD
102992-716
102992-718
102992-720
102992-722
LY2784544 ≥95%
LY2784544
LY2784544 is a potent JAK2 inhibitor with IC(50) of 3nM, effective in JAK2V617F, 8- and 20-fold selective versus JAK1 and JAK3. Phase 2.
Formula:
C₂₃H₂₅ClFN₇O MW: 469.9 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20482143 CAS Number: 1229236-86-5 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...