AZD-8330 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
2-((2-Fluoro-4-iodophenyl)amino)-N-(2-hydroxyethoxy)-1,5-dimethyl-6-oxo-1,6-dihydropyridine-3-carboxamide, AZD8330
SYN-1195-M001
SYN-1195-M005
SYN-1195-M010
SYN-1195-M050
102992-708EA
149.54
USD
102992-708
102992-710
102992-712
102992-714
AZD-8330 ≥95%
AZD-8330
AZD8330 is a novel, selective, non-ATP competitive MEK 1/2 inhibitor with an IC(50) of 7nM.
Formula:
C₁₆H₁₇FIN₃O₄ MW: 461.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18384989 CAS Number: 869357-68-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...