SGI-1776 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-((1-Methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine, SGI-1776 free base

SYN-1193-M001 SYN-1193-M005 SYN-1193-M010 SYN-1193-M050
102992-694EA 119.08 USD
102992-694 102992-696 102992-698 102992-700
SGI-1776 ≥95%
SGI-1776

SGI-1776 is a novel ATP competitive inhibitor of Pim1 with an IC(50) of 7nM, 50- and 10-fold selectivity versus Pim2 and Pim3. SGI-1776 inhibits FLT3 kinase as well, which can complicate some experimental results if not carefully controlled.

Formula: C₂₀H₂₂F₃N₅O
MW: 405.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16659064
CAS Number: 1025065-69-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR