XL388 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: (7-(6-Aminopyridin-3-yl)-2,3-dihydrobenzo[f][1,4]oxazepin-4(5H)-yl)(3-fluoro-2-methyl-4-(methylsulphonyl)phenyl)methanone

SYN-1186-M001 SYN-1186-M005 SYN-1186-M010 SYN-1186-M050
102992-646EA 231.23 USD
102992-646 102992-648 102992-650 102992-652
XL388 ≥95%
XL388

XL388 is a novel class of highly potent, selective, ATP-competitive, and orally bioavailable inhibitors of the mammalian target of rapamycin (mTOR) with an IC(50) of 9.9nM.

Formula: C₂₃H₂₂FN₃O₄S
MW: 455.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD24386875
CAS Number: 1251156-08-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR