CX-6258 ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
SYN-1182-M001
SYN-1182-M005
SYN-1182-M010
102992-620EA
283.85
USD
102992-620
102992-622
102992-624
CX-6258 ≥98% (by HPLC)
CX-6258
CX6258 is a potent inhibitor of Pim-1, Pim-2 and Pim-3 with IC(50) values of 5, 25 and 16nM, respectively.
Formula:
C₂₆H₂₄ClN₃O₃ Storage Temperature: Freezer |
CAS Number:
1202916-90-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...