PP242 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
SYN-1181-M001
SYN-1181-M005
SYN-1181-M010
SYN-1181-M050
102992-612EA
74.77
USD
102992-612
102992-614
102992-616
102992-618
PP242 ≥95%
PP242
PP242 exhibits potent selectivity for mTOR over other PI3K family kinases such as p110 alpha - delta and DNA-PK, with IC(50) of 1.96µM, 2.2µM, 1.27µM, 0.102µM, and 0.408µM, respectively.
Formula:
C₁₆H₁₆N₆O MW: 308.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12196869 CAS Number: 1092351-67-1 |
Specification Test Results
Purity | >95% |
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...