BGT226 maleate ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 8-(6-Methoxypyridin-3-yl)-3-methyl-1-(4-(piperazin-1-yl)-3-(trifluoromethyl)phenyl)-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one maleate, NVP-BEZ226

SYN-1178-M001 SYN-1178-M005 SYN-1178-M010 SYN-1178-M050
102992-588EA 134.31 USD
102992-588 102992-590 102992-592 102992-594
BGT226 maleate ≥95%
BGT226 maleate

NVP-BGT226 is a novel, orally bioavailable small-molecule inhibitor of phosphoinositol-3-kinase and mammalian target of rapamycin with an IC(50) of 1nM.

Formula: C₂₈H₂₅F₃N₆O₂·C₄H₄O₄
MW: 650.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22124895
CAS Number: 1245537-68-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR