SCH900776 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
6-Bromo-3-(1-methyl-1H-pyrazol-4-yl)-5-(3R)-3-piperidinylpyrazolo[1,5-a]pyrimidin-7-amine, MK-8776
SYN-1173-M001
SYN-1173-M005
SYN-1173-M010
SYN-1173-M050
102992-548EA
156.46
USD
102992-548
102992-550
102992-552
102992-554
SCH900776 ≥95%
SCH900776
SCH900776 is a selective Chk1 inhibitor with an IC(50) of 3nM. It shows 50-fold selectivity for Chk1 versus Chk2.
Formula:
C₁₅H₁₈BrN₇ MW: 376.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20922873 CAS Number: 891494-63-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...