LY-333,531 hydrochloride ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
(7S)-7-((Dimethylamino)methyl)-22,25-dihydro-11H,21H,31H-6-oxa-1,3(3,1)-diindola-2(3,4)-pyrrolacyclononaphane-22,25-dione hydrochloride, LY333531 hydrochloride
SYN-1172-M001
SYN-1172-M005
SYN-1172-M010
102992-542EA
246.46
USD
102992-542
102992-544
102992-546
LY-333,531 hydrochloride ≥95%
LY-333,531 hydrochloride
LY333531 is a potent, selective inhibitor of Protein Kinase C isozymes, PKCbetaI (IC(50) of 4.7nM) and PKCbetaII (IC(50) of 5.9nM).
Formula:
C₂₈H₂₈N₄O₃·HCl MW: 505.1 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00940226 CAS Number: 169939-93-9 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...