Ibrutinib ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: (R)-1-(3-(4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)prop-2-en-1-one, PCI-32765

SYN-1171-M001 SYN-1171-M005 SYN-1171-M010 SYN-1171-M050
102992-534EA 156.46 USD
102992-534 102992-536 102992-538 102992-540
Ibrutinib ≥95%
Ibrutinib

PCI-32765 (Ibrutinib) is a potent and highly selective Btk inhibitor with an IC(50) of 0.5nM.

Formula: C₂₅H₂₄N₆O₂
MW: 440.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20261150
CAS Number: 936563-96-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR