Regorafenib ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide, BAY-73-4506

SYN-1169-M001 SYN-1169-M005 SYN-1169-M010 SYN-1169-M050
102992-518EA 119.08 USD
102992-518 102992-520 102992-522 102992-524
Regorafenib ≥95%
Regorafenib

Regorafenib (BAY 73-4506, Fluoro-Sorafenib) is a multi-target inhibitor for VEGFR1, VEGFR2, VEGFR3, PDGFR, Kit, RET and Raf-1 with IC(50) values of 13, 4.2, 46, 22, 7, 1.5 and 2.5nM, respectively.

Formula: C₂₁H₁₅ClF₄N₄O₃
MW: 482.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16038047
CAS Number: 755037-03-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR