AZD-8055 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
AZD8055 is a novel ATP-competitive inhibitor of mTOR kinase activity with an IC(50) of 0.8nM. It shows excellent selectivity (~1,000-fold) against all class I phosphatidylinositol 3-kinase (PI3K) isoforms and other members of the PI3K-like kinase family.
Formula:
C₂₅H₃₁N₅O₄ Storage Temperature: Freezer |
MDL Number:
MFCD16660191 CAS Number: 1009298-09-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...