GCI1746 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: G-182, 4-(tert-Butyl)-N-(2-methyl-3-(4-methyl-6-((4-(morpholine-4-carbonyl)phenyl)amino)-5-oxo-4,5-dihydropyrazin-2-yl)phenyl)benzamide, CGI-1746

SYN-1164-M001 SYN-1164-M005 SYN-1164-M010
102992-484EA 246.46 USD
102992-484 102992-486 102992-488
GCI1746 ≥95%
GCI1746

CGI1746 was identified as a selective and potent ATP-competitive BTK inhibitor with an IC(50) of 1.9nM. It is specific for BTK, with around 1000-fold selectivity over Tec and Src family kinases.

Formula: C₃₄H₃₇N₅O₄
MW: 579.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18782602
CAS Number: 910232-84-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR