MK-2206 dihydrochloride ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1162-M001 SYN-1162-M005 SYN-1162-M010 SYN-1162-M050
102992-470EA 105.23 USD
102992-470 102992-472 102992-474 102992-476
MK-2206 dihydrochloride ≥95%
MK-2206 dihydrochloride

MK-2206 is a highly selective inhibitor of Akt1, Akt2 and Akt3 with IC(50) of 8nM, 12nM and 65nM, respectively.

Formula: C₂₅H₂₁N₅O·2HCl
Storage Temperature: Freezer
MDL Number: MFCD14584463
CAS Number: 1032350-13-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR