PLX4032 ≥95%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: N-(3-(5-(4-Chlorophenyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulphonamide, Vemurafenib

SYN-1161-M001 SYN-1161-M005 SYN-1161-M010 SYN-1161-M050
102992-462EA 74.77 USD
102992-462 102992-464 102992-466 102992-468
PLX4032 ≥95%
PLX4032

PLX4032 (Vemurafenib, RG7204, Zelboraf, RO5185426) is a novel and potent inhibitor of B-RafV600E with an IC(50) of 31nM.

Formula: C₂₃H₁₈ClF₂N₃O₃S
MW: 489.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18074504
CAS Number: 918504-65-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR