TAE684 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 5-Chloro-N4-(2-(isopropylsulphonyl)phenyl)-N2-(2-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)pyrimidine-2,4-diamine, NVP-TAE 684, NVP-TAE684

SYN-1159-M001 SYN-1159-M005 SYN-1159-M010 SYN-1159-M050
102992-448EA 134.31 USD
102992-448 102992-450 102992-452 102992-454
TAE684 ≥95%
TAE684

TAE684 is a potent and selective ALK inhibitor with an IC(50) of 3nM. It is 100-fold more sensitive for ALK over InsR.

Formula: C₃₀H₄₀ClN₇O₃S
MW: 614.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11977634
CAS Number: 761439-42-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR