MLN0128 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1157-M001 SYN-1157-M005 SYN-1157-M010 SYN-1157-M050
102992-434EA 142.62 USD
102992-434 102992-436 102992-438 102992-440
MLN0128 ≥95%
MLN0128

MLN0128 is a potent and selective mTOR inhibitor with an IC(50) of 1nM; 200-fold less potent to class I PI3K isoforms, superior in blocking mTORC1/2 and sensitive to pro-invasion genes (vs Rapamycin).

Formula: C₁₅H₁₅N₇O
Storage Temperature: Freezer
MDL Number: MFCD22124893
CAS Number: 1224844-38-5

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR