MK-2461 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1149-M001 SYN-1149-M005 SYN-1149-M010
102992-378EA 246.46 USD
102992-378 102992-380 102992-382
MK-2461 ≥95%
MK-2461

MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met kinases (N1100Y, Y1230C, Y1230H, Y1235D, and M1250T) with IC(50) values of 0.4 to 2.5nM.

Formula: C₂₄H₂₅N₅O₅S
Storage Temperature: Freezer
MDL Number: MFCD11977739
CAS Number: 917879-39-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR