GNE-477 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
5-(7-Methyl-6-((4-(methylsulphonyl)piperazin-1-yl)methyl)-4-morpholinothieno[3,2-d]pyrimidin-2-yl)pyrimidin-2-amine
SYN-1148-M001
SYN-1148-M005
SYN-1148-M010
102992-372EA
292.15
USD
102992-372
102992-374
102992-376
GNE-477 ≥95%
GNE-477
GNE 477 is a potent and efficacious dual PI3K/mTOR inhibitor with an IC(50) value of 4nM and 21nM respectively.
Formula:
C₂₁H₂₈N₈O₃S₂ MW: 504.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20527726 CAS Number: 1032754-81-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...