CH5132799 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
5-(7-(Methylsulphonyl)-2-morpholino-6,7-dihydro-5H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrimidin-2-amine, CH-5132799, Izorlisib
SYN-1146-M001
SYN-1146-M005
SYN-1146-M010
SYN-1146-M050
102992-358EA
186.92
USD
102992-358
102992-360
102992-362
102992-364
CH5132799 ≥95%
CH5132799
CH5132799 selectively inhibits class I PI3Kalpha with an IC(50) value of 14nM.
Formula:
C₁₅H₁₉N₇O₃S MW: 377.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD22419020 CAS Number: 1007207-67-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...