NIBR-17 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
N6-(6-Methoxypyridin-3-yl)-2-morpholino-[4,5'-bipyrimidine]-2',6-diamine
SYN-1145-M001
SYN-1145-M005
SYN-1145-M010
SYN-1145-M050
102992-350EA
254.77
USD
102992-350
102992-352
102992-354
102992-356
NIBR-17 ≥95%
NIBR-17
NIBR-17 is a pan class I PI3K inhibitor with IC(50) values of 1, 9.2, 9 and 20nM, respectively for PI3Kalpha - delta.
Formula:
C₁₈H₂₀N₈O₂ MW: 380.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD22631503 CAS Number: 944396-88-7 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...