AMG-1 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1143-M001 SYN-1143-M005 SYN-1143-M010
102992-336EA 246.46 USD
102992-336 102992-338 102992-340
AMG-1 ≥95%
AMG-1

AMG-1 is a potent inhibitor of human c-Met and RON with IC(50) values of 4 and 9nM respectively.

Formula: C₃₁H₂₉FN₄O₅
Storage Temperature: Freezer
MDL Number: MFCD22631502
CAS Number: 913376-84-8

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR