Neratinib ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: HKI-272, (E)-N-(4-((3-Chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide

SYN-1140-M001 SYN-1140-M005 SYN-1140-M010 SYN-1140-M050
102992-312EA 119.08 USD
102992-312 102992-314 102992-316 102992-318
Neratinib ≥95%
Neratinib

Neratinib (HKI-272) is an orally available, irreversible tyrosine kinase inhibitor with IC(50) values of 59nM and 92nM for HER2 and EGFR, respectively. It is in development for the treatment of early- and late-stage HER2-positive breast cancer.

Formula: C₃₀H₂₉ClN₆O₃
Storage Temperature: Freezer
MDL Number: MFCD09752958
CAS Number: 698387-09-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR