Cabozantinib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL184
SYN-1138-M001
SYN-1138-M005
SYN-1138-M010
SYN-1138-M050
102992-304EA
96.92
USD
102992-304
102992-306
102992-308
102992-310
Cabozantinib ≥95%
Cabozantinib
Cabozantinib is very potent inhibitor, with an IC(50) of less than 10nM for VEGFR2, MET, Kit and FLT3-ITD.
Formula:
C₂₈H₂₄FN₃O₅ MW: 501.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20926324 CAS Number: 849217-68-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...